The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020471.1 | NP_065204.1 | 1 - 2 | 135 | 102 |
XM_006243089.1 | XP_006243151.1 | 1 - 2 | 294 | 102 | |
GenBank mRNA | L35571.1 | - | 1 - 2 | 135 | 102 |