The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021664.1 | NP_067696.1 | 1 - 2 | 270 | 81 |
XM_006233464.2 | XP_006233526.1 | 2 - 3 | 347 | 81 | |
XM_006233465.2 | XP_006233527.1 | 2 - 3 | 374 | 81 | |
XM_006233466.2 | XP_006233528.1 | 2 - 3 | 377 | 81 | |
XM_006233468.2 | XP_006233530.1 | 2 - 3 | 758 | 81 | |
XM_008761521.1 | XP_008759743.1 | 2 - 3 | 480 | 81 | |
GenBank mRNA | AF178974.1 | - | 1 - 2 | 270 | 81 |