Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slc26a1
- Entrez Gene ID
-
64076
- Gene Name
- solute carrier family 26 (anion exchanger), member 1
- Gene Aliases
-
sat-1
- Chromosome Location
-
Chr.14: 2050805 - 2056091 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
L23413.1 |
- |
1 - 2 |
338 |
67 |
View Full Product Details