The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022512.2 | NP_071957.1 | 2 - 3 | 241 | 87 |
GenBank mRNA | BC072545.1 | - | 2 - 3 | 260 | 87 |
J05030.1 | - | 2 - 3 | 242 | 87 |