Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hand2
Entrez Gene ID
64637
Gene Name
heart and neural crest derivatives expressed 2
Gene Aliases
-
Chromosome Location
Chr.16: 36370715 - 36373634 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022696.1 NP_073187.1 - 587 66
GenBank mRNA Y08138.1 - - 587 66

View Full Product Details