The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033870.1 | NP_001029042.1 | 2 - 3 | 646 | 81 |
NM_023102.2 | NP_075590.2 | 2 - 3 | 646 | 81 | |
XM_006241000.2 | XP_006241062.1 | 2 - 3 | 1943 | 81 | |
XM_006241001.2 | XP_006241063.1 | 2 - 3 | 651 | 81 | |
XM_006241002.2 | XP_006241064.1 | 2 - 3 | 1941 | 81 | |
XM_008765130.1 | XP_008763352.1 | 2 - 3 | 651 | 81 | |
GenBank mRNA | BC072533.1 | - | 2 - 3 | 646 | 81 |
U22297.1 | - | 1 - 2 | 81 |