The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031522.3 | NP_113710.2 | 3 - 4 | 776 | 79 |
GenBank mRNA | AB035722.1 | - | 3 - 4 | 634 | 79 |
BC100639.1 | - | 3 - 4 | 651 | 79 |