The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031595.1 | NP_113783.1 | 8 - 9 | 1121 | 87 |
GenBank mRNA | BC062019.1 | - | 8 - 9 | 1031 | 87 |
D83522.1 | - | 8 - 9 | 995 | 87 | |
FQ225567.1 | - | 7 - 8 | 780 | 87 | |
FQ225947.1 | - | 7 - 8 | 780 | 87 | |
FQ226273.1 | - | 7 - 8 | 780 | 87 | |
FQ232987.1 | - | 7 - 8 | 781 | 87 | |
U77918.1 | - | 8 - 9 | 1121 | 87 |