The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032079.2 | NP_114468.2 | 6 - 7 | 795 | 120 |
GenBank mRNA | BC087010.1 | - | 6 - 7 | 795 | 120 |
FQ231743.1 | - | 4 - 5 | 551 | 120 | |
U95727.1 | - | 6 - 7 | 829 | 120 |