The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053388.1 | NP_445840.1 | 2 - 3 | 253 | 81 |
XM_008770784.1 | XP_008769006.1 | 2 - 3 | 364 | 81 | |
GenBank mRNA | AF170284.1 | - | 2 - 3 | 253 | 81 |