The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053552.1 | NP_446004.1 | 2 - 3 | 367 | 101 |
GenBank mRNA | AF037067.1 | - | 2 - 3 | 367 | 101 |