Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ntn1
Entrez Gene ID
114523
Gene Name
netrin 1
Gene Aliases
netrin-1
Chromosome Location
Chr.10: 54761925 - 54982039 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053731.2 NP_446183.2 2 - 3 1212 93
XM_006246567.2 XP_006246629.1 3 - 4 1564 93
XM_006246568.2 XP_006246630.1 3 - 4 1398 93
GenBank mRNA AY028417.1 - 2 - 3 1212 93

View Full Product Details