Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map3k8
Entrez Gene ID
116596
Gene Name
mitogen-activated protein kinase kinase kinase 8
Gene Aliases
D17TPL2, Tpl-2, Tpl2
Chromosome Location
Chr.17: 56109403 - 56129925 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053847.1 NP_446299.1 1 - 2 298 89
GenBank mRNA FQ217924.1 - 1 - 2 331 89
M94454.1 - 1 - 2 298 89

View Full Product Details