The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053965.2 | NP_446417.2 | 7 - 8 | 822 | 80 |
GenBank mRNA | BC081749.1 | - | 7 - 8 | 822 | 80 |
FQ209681.1 | - | 7 - 8 | 778 | 80 | |
X97831.1 | - | 7 - 8 | 779 | 80 |