The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053978.1 | NP_446430.1 | 6 - 7 | 587 | 96 |
XM_008770197.1 | XP_008768419.1 | 6 - 7 | 1165 | 96 | |
GenBank mRNA | X78606.1 | - | 6 - 7 | 587 | 96 |