Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- P2ry6
- Entrez Gene ID
-
117264
- Gene Name
- pyrimidinergic receptor P2Y, G-protein coupled, 6
- Gene Aliases
-
P2y6
- Chromosome Location
-
Chr.1: 165972439 - 166008351 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
D63665.1 |
- |
1 - 2 |
69 |
127 |
View Full Product Details