The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057188.1 | NP_476536.1 | 5 - 6 | 621 | 99 |
GenBank mRNA | AF090867.1 | - | 5 - 6 | 621 | 99 |