The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057206.1 | NP_476554.1 | 9 - 10 | 1181 | 93 |
GenBank mRNA | L09216.1 | - | 9 - 10 | 1181 | 93 |