The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133299.2 | NP_579833.1 | 4 - 5 | 604 | 81 |
XM_008767180.1 | XP_008765402.1 | 4 - 5 | 610 | 81 | |
GenBank mRNA | AF021854.1 | - | 4 - 5 | 605 | 81 |
AF099742.1 | - | 4 - 5 | 524 | 81 | |
BC060546.1 | - | 4 - 5 | 598 | 81 | |
FQ209650.1 | - | 1 - 2 | 81 | 81 | |
FQ210848.1 | - | 1 - 2 | 81 | 81 | |
FQ218655.1 | - | 1 - 2 | 81 | 81 | |
FQ218673.1 | - | 1 - 2 | 81 | 81 | |
FQ218683.1 | - | 1 - 2 | 81 | 81 | |
FQ218690.1 | - | 1 - 2 | 81 | 81 | |
FQ219508.1 | - | 1 - 2 | 81 | 81 |