Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mmp10
Entrez Gene ID
117061
Gene Name
matrix metallopeptidase 10 (stromelysin 2)
Gene Aliases
-
Chromosome Location
Chr.8: 5734348 - 5742243 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133514.1 NP_598198.1 6 - 7 968 79
GenBank mRNA M65253.1 - 6 - 7 968 79
X05083.1 - 6 - 7 965 79

View Full Product Details