Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Svs5
Entrez Gene ID
171027
Gene Name
seminal vesicle secretory protein 5
Gene Aliases
-
Chromosome Location
Chr.3: 160713717 - 160715327 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133516.2 NP_598200.1 2 - 3 436 107
GenBank mRNA AF159097.1 - 1 - 2 205 107
DQ232687.1 - 1 - 2 52 107
X01115.1 - 2 - 3 436 106

Gene Symbol
LOC103694892
Entrez Gene ID
103694892
Gene Name
seminal vesicle secretory protein 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_007906879: 62519 - 64164 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008773873.1 XP_008772095.1 2 - 3 471 107

View Full Product Details