The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133566.1 | NP_598250.1 | 1 - 2 | 517 | 78 |
GenBank mRNA | AY046415.1 | - | 1 - 2 | 517 | 78 |