The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138848.1 | NP_620203.1 | 4 - 5 | 1032 | 75 |
XM_008762758.1 | XP_008760980.1 | 4 - 5 | 1192 | 75 | |
GenBank mRNA | AB020726.1 | - | 4 - 5 | 1105 | 75 |
AF109393.1 | - | 4 - 5 | 1032 | 75 | |
BC070886.1 | - | 4 - 5 | 1102 | 75 |