Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tor1aip1
- Entrez Gene ID
-
246314
- Gene Name
- torsin A interacting protein 1
- Gene Aliases
-
Lap1b, Lap1c
- Chromosome Location
-
Chr.13: 73670649 - 73704668 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U19614.1 |
- |
3 - 4 |
675 |
123 |
View Full Product Details