The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164073.1 | NP_001157545.1 | 7 - 8 | 1197 | 112 |
NM_145788.2 | NP_665731.2 | 7 - 8 | 1208 | 112 | |
XM_006234273.2 | XP_006234335.1 | 7 - 8 | 1270 | 112 | |
XM_006234274.2 | XP_006234336.1 | 7 - 8 | 1256 | 112 | |
GenBank mRNA | AJ422211.1 | - | 6 - 7 | 1068 | 112 |
BC088854.1 | - | 8 - 9 | 1276 | 112 | |
BC129086.1 | - | 7 - 8 | 1197 | 112 |