The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147165.1 | NP_671694.1 | 2 - 3 | 288 | 76 |
GenBank mRNA | M76733.1 | - | 2 - 3 | 288 | 76 |