The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_171990.2 | NP_741987.1 | 6 - 7 | 551 | 79 |
XM_006250431.2 | XP_006250493.1 | 6 - 7 | 647 | 79 | |
GenBank mRNA | AJ000347.1 | - | 6 - 7 | 502 | 79 |
BC085692.1 | - | 6 - 7 | 551 | 79 |