Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mrgprx1
- Entrez Gene ID
-
282547
- Gene Name
- MAS-related GPR, member X1
- Gene Aliases
-
Mgrg2, Mrgprc11, Snsr1
- Chromosome Location
-
Chr.1: 103379325 - 103380338 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_172158.2 |
NP_750845.2 |
1 - 1 |
744 |
77 |
- Gene Symbol
- Mrgprc
- Entrez Gene ID
-
404646
- Gene Name
- MAS-related G-protein coupled receptor, member C
- Gene Aliases
-
MrgC, Snsr
- Chromosome Location
-
-
View Full Product Details