Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppargc1b
Entrez Gene ID
291567
Gene Name
peroxisome proliferator-activated receptor gamma, coactivator 1 beta
Gene Aliases
PGC1beta, Perc
Chromosome Location
Chr.18: 56626725 - 56650524 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176075.2 NP_788264.1 - 402 54
GenBank mRNA AY188951.1 - - 402 54

View Full Product Details