The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107959.1 | NP_001101429.1 | 7 - 8 | 910 | 59 |
XM_006238656.2 | XP_006238718.1 | 7 - 8 | 1726 | 59 |