The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021594.1 | NP_067605.1 | 5 - 6 | 951 | 64 |
GenBank mRNA | AF154336.1 | - | 5 - 6 | 951 | 64 |
FQ213792.1 | - | 5 - 6 | 1068 | 64 | |
FQ228926.1 | - | 5 - 6 | 1049 | 64 |