Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Flot1
Entrez Gene ID
64665
Gene Name
flotillin 1
Gene Aliases
Rareg
Chromosome Location
Chr.20: 3427890 - 3438409 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022701.2 NP_073192.2 4 - 5 385 120
XM_006256024.2 XP_006256086.1 4 - 5 404 120
GenBank mRNA BC064652.1 - 4 - 5 384 120
CK359020.1 - 4 - 5 385 120
U60976.1 - 4 - 5 315 120
U60977.1 - 3 - 4 389 120

View Full Product Details