Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nolc1
Entrez Gene ID
64896
Gene Name
nucleolar and coiled-body phosphoprotein 1
Gene Aliases
Nopp140
Chromosome Location
Chr.1: 265829057 - 265839819 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022869.1 NP_074060.1 12 - 13 2006 96
XM_006231560.2 XP_006231622.1 12 - 13 2054 96
GenBank mRNA M94287.1 - 12 - 13 2006 96

View Full Product Details