Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc6a1
Entrez Gene ID
79212
Gene Name
solute carrier family 6 (neurotransmitter transporter), member 1
Gene Aliases
Gabt1, Gat1
Chromosome Location
Chr.4: 146258834 - 146292176 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024371.2 NP_077347.1 6 - 7 857 89
XM_006237059.1 XP_006237121.1 7 - 8 1066 89
XM_006237060.2 XP_006237122.1 7 - 8 3286 89
GenBank mRNA M59742.1 - 6 - 7 857 89

View Full Product Details