The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031000.3 | NP_112262.1 | 3 - 4 | 259 | 72 |
GenBank mRNA | BC059133.1 | - | 3 - 4 | 538 | 72 |
D10854.1 | - | 3 - 4 | 259 | 72 | |
FQ212523.1 | - | 3 - 4 | 501 | 72 | |
FQ212586.1 | - | 3 - 4 | 499 | 72 | |
FQ212779.1 | - | 3 - 4 | 534 | 72 | |
FQ212919.1 | - | 2 - 3 | 209 | 72 | |
FQ213220.1 | - | 2 - 3 | 206 | 72 | |
FQ213467.1 | - | 3 - 4 | 534 | 72 | |
FQ213483.1 | - | 3 - 4 | 530 | 72 | |
FQ213491.1 | - | 3 - 4 | 532 | 72 | |
FQ213579.1 | - | 3 - 4 | 501 | 72 | |
FQ213776.1 | - | 3 - 4 | 501 | 72 | |
FQ214173.1 | - | 3 - 4 | 501 | 72 | |
FQ214441.1 | - | 3 - 4 | 278 | 72 | |
FQ214642.1 | - | - | 516 | 72 | |
FQ214700.1 | - | 3 - 4 | 474 | 72 | |
FQ215620.1 | - | 3 - 4 | 326 | 72 | |
FQ215772.1 | - | 3 - 4 | 501 | 72 | |
FQ218383.1 | - | 3 - 4 | 501 | 72 | |
FQ218721.1 | - | 3 - 4 | 501 | 72 | |
FQ218729.1 | - | 3 - 4 | 501 | 72 | |
FQ220055.1 | - | 1 - 2 | 102 | 72 | |
FQ226221.1 | - | 3 - 4 | 534 | 72 | |
FQ228131.1 | - | 3 - 4 | 534 | 72 | |
FQ228132.1 | - | 3 - 4 | 534 | 72 | |
FQ228842.1 | - | 3 - 4 | 530 | 72 |