The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031316.1 | NP_112606.1 | 3 - 4 | 319 | 72 |
GenBank mRNA | AB019119.1 | - | 3 - 4 | 319 | 72 |
AF150741.1 | - | 3 - 4 | 319 | 72 |