Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scamp4
Entrez Gene ID
65170
Gene Name
secretory carrier membrane protein 4
Gene Aliases
-
Chromosome Location
Chr.7: 11969720 - 11982141 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031725.1 NP_113913.1 4 - 5 369 75
XM_006240999.1 XP_006241061.1 4 - 5 388 75
GenBank mRNA AF285631.1 - 4 - 5 369 75
BC090342.1 - 4 - 5 363 75

View Full Product Details