The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053937.1 | NP_446389.1 | 12 - 13 | 2600 | 85 |
XM_006247554.2 | XP_006247616.1 | 13 - 14 | 2691 | 85 | |
XM_006247555.2 | XP_006247617.1 | 13 - 14 | 2649 | 85 | |
XM_006247556.2 | XP_006247618.1 | 12 - 13 | 2628 | 85 | |
GenBank mRNA | AF016192.1 | - | 12 - 13 | 2600 | 85 |