Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a20
Entrez Gene ID
117035
Gene Name
solute carrier family 25 (carnitine/acylcarnitine translocase), member 20
Gene Aliases
-
Chromosome Location
Chr.8: 117455308 - 117476762 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053965.2 NP_446417.2 4 - 5 520 84
GenBank mRNA BC081749.1 - 4 - 5 520 84
FQ209681.1 - 4 - 5 476 84
X97831.1 - 4 - 5 477 84

View Full Product Details