Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Qtrt1
Entrez Gene ID
64016
Gene Name
queuine tRNA-ribosyltransferase 1
Gene Aliases
Tgt, Tgut
Chromosome Location
Chr.8: 22447057 - 22454328 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022250.2 NP_071586.2 4 - 5 540 81
GenBank mRNA AB034634.1 - 4 - 5 535 81
BC097321.1 - 4 - 5 540 81

View Full Product Details