The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022250.2 | NP_071586.2 | 5 - 6 | 661 | 60 |
GenBank mRNA | AB034634.1 | - | 5 - 6 | 656 | 60 |
BC097321.1 | - | 5 - 6 | 661 | 60 |