Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fgf20
Entrez Gene ID
66017
Gene Name
fibroblast growth factor 20
Gene Aliases
-
Chromosome Location
Chr.16: 55152748 - 55159352 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023961.1 NP_076451.1 2 - 3 388 131
GenBank mRNA AB020021.1 - 2 - 3 388 131

View Full Product Details