Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Becn1
Entrez Gene ID
114558
Gene Name
beclin 1, autophagy related
Gene Aliases
-
Chromosome Location
Chr.10: 89209944 - 89225297 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034117.1 NP_001029289.1 3 - 4 398 59
NM_053739.2 NP_446191.1 3 - 4 420 59
GenBank mRNA AY033824.1 - 2 - 3 196 59
BC074011.1 - 3 - 4 285 59
CK600131.1 - 3 - 4 398 59

View Full Product Details