Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc36a1
Entrez Gene ID
155205
Gene Name
solute carrier family 36 (proton/amino acid symporter), member 1
Gene Aliases
Lyaat1
Chromosome Location
Chr.10: 40538013 - 40573304 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130415.1 NP_569099.1 2 - 3 298 65
XM_008767657.1 XP_008765879.1 1 - 2 2397 65
XM_008767659.1 XP_008765881.1 3 - 4 980 65
GenBank mRNA AF361239.1 - 2 - 3 298 65

View Full Product Details