Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcsk2
Entrez Gene ID
25121
Gene Name
proprotein convertase subtilisin/kexin type 2
Gene Aliases
-
Chromosome Location
Chr.3: 137618891 - 137923385 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012746.1 NP_036878.1 6 - 7 940 68
GenBank mRNA M76706.1 - 6 - 7 940 68
M83746.1 - 6 - 7 912 68

View Full Product Details