The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013113.2 | NP_037245.2 | 5 - 6 | 789 | 114 |
GenBank mRNA | BC078902.1 | - | 5 - 6 | 789 | 114 |
J02701.1 | - | 5 - 6 | 1112 | 114 | |
M14137.1 | - | 5 - 6 | 693 | 114 |