Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ndel1
Entrez Gene ID
170845
Gene Name
nudE neurodevelopment protein 1-like 1
Gene Aliases
NUDE2, Nudel
Chromosome Location
Chr.10: 55411888 - 55456654 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133320.1 NP_579854.1 4 - 5 452 86
XM_006246571.2 XP_006246633.1 4 - 5 861 86
XM_006246572.2 XP_006246634.1 4 - 5 854 86
XM_006246573.2 XP_006246635.1 4 - 5 602 86
XM_006246574.2 XP_006246636.1 4 - 5 487 86
XM_006246575.2 XP_006246637.1 4 - 5 965 86
XM_006246576.2 XP_006246638.1 4 - 5 855 86
XM_006246578.2 XP_006246640.1 4 - 5 601 86
GenBank mRNA AY008298.1 - 4 - 5 452 86
BC070909.1 - 4 - 5 542 86

View Full Product Details