Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep170
Entrez Gene ID
171457
Gene Name
centrosomal protein 170
Gene Aliases
KAB1, Kab, Rhk1
Chromosome Location
Chr.13: 94807090 - 94887448 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB038495.1 - - 211 74

Gene Symbol
LOC100909664
Entrez Gene ID
100909664
Gene Name
centrosomal protein of 170 kDa-like
Gene Aliases
-
Chromosome Location
Chr.13: 43806604 - 43845827 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751234.3 XP_003751282.2 2 - 3 1431 74
XM_006249680.2 XP_006249742.1 2 - 3 1432 74
XM_006249681.2 XP_006249743.1 2 - 3 1432 74
XM_006249682.2 XP_006249744.1 2 - 3 1431 74
XM_006249683.2 XP_006249745.1 2 - 3 1431 74

View Full Product Details