Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tank
Entrez Gene ID
252961
Gene Name
TRAF family member-associated NFKB activator
Gene Aliases
-
Chromosome Location
Chr.3: 47439006 - 47515706 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164073.1 NP_001157545.1 3 - 4 335 78
NM_145788.2 NP_665731.2 3 - 4 346 78
XM_006234273.2 XP_006234335.1 3 - 4 408 78
XM_006234274.2 XP_006234336.1 3 - 4 394 78
GenBank mRNA AJ422211.1 - 2 - 3 206 78
BC088854.1 - 3 - 4 283 78
BC129086.1 - 3 - 4 335 78
EV776506.1 - 3 - 4 346 78

View Full Product Details