Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fgl1
Entrez Gene ID
246186
Gene Name
fibrinogen-like 1
Gene Aliases
Frep1, Lfire1
Chromosome Location
Chr.16: 54153086 - 54188120 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172010.1 NP_742007.1 2 - 3 161 73
GenBank mRNA AF508020.1 - 2 - 3 161 73
BC088107.1 - 2 - 3 117 73

View Full Product Details